2-Acetylamino-4-methylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

M2787-1G M2787-5G
TCM2787-1GEA 42.66 CAD
TCM2787-1G TCM2787-5G
2-Acetylamino-4-methylpyridine ≥98.0% (by GC)
2-Acetylamino-4-methylpyridine
Formula: C₈H₁₀N₂O
MDL Number: MFCD00234432
CAS Number: 5327-32-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 106.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR