1-Benzoylpiperidine ≥98.0% (by GC)
Supplier: TCI America
B5512-1G
B5512-5G
76219-638EA
66.83
CAD
76219-638
76219-640
1-Benzoylpiperidine ≥98.0% (by GC)
1-Benzoylpiperidine
Formula:
C₁₂H₁₅NO Boiling Pt: 321°C |
MDL Number:
MFCD00023702 CAS Number: 776-75-0 EINECS: 212-280-3 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 48.0 to 52.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...