5-Cyano-2-methylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 6-Methylnicotinonitrile, 5-Cyano-2-picoline

C3050-1G C3050-5G
TCC3050-1GEA 74.26 CAD
TCC3050-1G TCC3050-5G
5-Cyano-2-methylpyridine ≥98.0% (by GC, titration analysis)
5-Cyano-2-methylpyridine
Formula: C₇H₆N₂
Boiling Pt: 217°C
MDL Number: MFCD00038042
CAS Number: 3222-48-8
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR