3-Nitro-4-quinolinol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

N0992-1G N0992-200MG
TCN0992-1GEA 581.76 CAD
TCN0992-1G TCN0992-200MG
3-Nitro-4-quinolinol ≥98.0% (by GC, titration analysis)
3-Nitro-4-quinolinol
Formula: C₉H₆N₂O₃
Melting Pt: 361°C
MDL Number: MFCD00277722
CAS Number: 50332-66-6

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR