3-Cyano-4-methylaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 5-Amino-2-methylbenzonitrile

A2814-1G A2814-5G
TCA2814-1GEA 26.86 CAD
TCA2814-1G TCA2814-5G
3-Cyano-4-methylaniline ≥98.0% (by GC, titration analysis)
3-Cyano-4-methylaniline
Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 110°C
MDL Number: MFCD00017608
CAS Number: 50670-64-9
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 89.0 to 93.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR