6-Iodo-m-toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

I1034-1G I1034-5G
TCI1034-1GEA 55.3 CAD
TCI1034-1G TCI1034-5G
6-Iodo-m-toluidine ≥98.0% (by GC, titration analysis)
6-Iodo-m-toluidine
Formula: C₇H₈IN
Storage Temperature: Refrigerator
MDL Number: MFCD00833395
CAS Number: 13194-69-9

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 35.0 to 39.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR