5-Hydroxypicolinic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
H1471-1G
H1471-5G
TCH1471-1GEA
164.32
CAD
TCH1471-1G
TCH1471-5G
5-Hydroxypicolinic acid ≥98.0% (by HPLC, titration analysis)
5-Hydroxypicolinic acid
Formula:
C₆H₅NO₃ Melting Pt: 268°C |
MDL Number:
MFCD00661309 CAS Number: 15069-92-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...