4-Amino-2,6-dimethylpyridine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
A2749-1G
A2749-5G
TCA2749-1GEA
167.48
CAD
TCA2749-1G
TCA2749-5G
4-Amino-2,6-dimethylpyridine ≥98.0% (by GC)
4-Amino-2,6-dimethylpyridine
Formula:
C₇H₁₀N₂ Boiling Pt: 246°C |
MDL Number:
MFCD00130078 CAS Number: 3512-80-9 EINECS: 222-515-1 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 191.0 to 195.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...