5-Iodo-2-methoxypyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

I0925-1G I0925-5G
TCI0925-1GEA 62.88 CAD
TCI0925-1G TCI0925-5G
5-Iodo-2-methoxypyridine ≥98.0% (by GC)
5-Iodo-2-methoxypyridine
Formula: C₆H₆INO
Boiling Pt: 74°C
MDL Number: MFCD07781180
CAS Number: 13472-61-2

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR