1-(4-Chlorophenyl)cyclopentanecarboxylic acid 98,0 GC, T

Supplier: TCI America

Synonyms: 1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid

C2912-25G C2912-5G
TCC2912-25GEA 301.78 CAD
TCC2912-25G TCC2912-5G
1-(4-Chlorophenyl)cyclopentanecarboxylic acid 98,0 GC, T
1-(4-Chlorophenyl)cyclopentanecarboxylic acid
Formula: C₁₂H₁₃ClO₂
MW: 224.6834 g/mol
MDL Number: MFCD00001373
CAS Number: 80789-69-1
EINECS: 279-552-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 162.0 to 166.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR