3-Nitro-2-picoline ≥98.0% (by GC)

Supplier: TCI America
Warning

M2548-1G M2548-5G
TCM2548-1GEA 50.56 CAD
TCM2548-1G TCM2548-5G
3-Nitro-2-picoline ≥98.0% (by GC)
3-Nitro-2-picoline
Formula: C₆H₆N₂O₂
Boiling Pt: 86°C
Storage Temperature: Refrigerator
MDL Number: MFCD04114137
CAS Number: 18699-87-1

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR