5-Acetyl-2-chloropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Acetyl-6-chloropyridine

A2796-1G A2796-5G
TCA2796-1GEA 64.78 CAD
TCA2796-1G TCA2796-5G
5-Acetyl-2-chloropyridine ≥98.0% (by GC)
5-Acetyl-2-chloropyridine
Formula: C₇H₆ClNO
MW: 155.5816 g/mol
MDL Number: MFCD03407343
CAS Number: 55676-22-7

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 100.0 to 104.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR