Thermal Transfer Printer Ribbons, Brady®
Supplier: Brady
R4306 R4300 IP-R4302
CA94000-380EA 258.63 CAD
CA94000-380 CA89066-904 CA89077-376
Thermal Transfer Printer Ribbons, Brady®
Printer Ribbons
A variety of ribbons designed for high-performance printing on Brady thermal transfer printing systems.
  • Reliable and sharp-edge print quality helps to maximize bar code readability
The 4300 series provides strong environmental resistance against smearing and chemicals.
The high density 6000 and 6400 series provide excellent solvent resistance, high heat resistance, and superior scratch resistance.
Delivery information: All Brady IP™ Thermal Transfer printer enabled ribbons include a spindle. For additional information on Brady IP™ printers, see 89077-382.