5-Acetyluracil ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5-Acetylpyrimidine-2,4(1H,3H)-dione

A2670-1G A2670-5G
TCA2670-1GEA 94.8 CAD
TCA2670-1G TCA2670-5G
5-Acetyluracil ≥98.0% (by HPLC, titration analysis)
5-Acetyluracil
Formula: C₆H₆N₂O₃
MW: 154.11 g/mol
MDL Number: MFCD00151968
CAS Number: 6214-65-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR