1-Acetylisatin ≥98.0% (by GC)

Supplier: TCI America
Warning

A2941-25G A2941-5G
76218-758EA 769.46 CAD
76218-758 76218-760
1-Acetylisatin ≥98.0% (by GC)
1-Acetylisatin
Formula: C₁₀H₇NO₃
MDL Number: MFCD00158542
CAS Number: 574-17-4
EINECS: 209-368-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR