(S)-1-Boc-2-methylpiperazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: (S)-2-Methyl-1-piperazinecarboxylic acid tert-Butyl ester, tert-Butyl (S)-2-Methyl-1-piperazinecarboxylate, (S)-tert-Butyl 2-methylpiperazine-1-carboxylate, (S)-1-Boc-2-methylpiperazine, (S)-1-(tert-Butoxycarbonyl)-2-methylpiperazine

B4846-1G B4846-5G
TCB4846-1GEA 95.91 CAD
TCB4846-1G TCB4846-5G
(S)-1-Boc-2-methylpiperazine ≥98.0% (by GC, titration analysis)
(S)-1-Boc-2-methylpiperazine
Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD01862121
CAS Number: 169447-70-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 43.0 to 47.0 deg-C
Specific rotation [a]20/D +68.0 to +72.0 deg (C=0.65, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR