1-Boc-piperidine ≥98.0% (by GC)

Supplier: TCI America
Warning

B4625-25G B4625-5G
TCB4625-25GEA 237.6 CAD
TCB4625-25G TCB4625-5G
1-Boc-piperidine ≥98.0% (by GC)
1-Boc-piperidine
Formula: C₁₀H₁₉NO₂
Boiling Pt: 120°C
Flash Pt: 86°C
MDL Number: MFCD02093935
CAS Number: 75844-69-8

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid to slightly cloudy liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR