(R)-(-)-3-Aminopiperidine dihydrochloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A2787-1G A2787-200MG
TCA2787-1GEA 134.3 CAD
TCA2787-1G TCA2787-200MG
(R)-(-)-3-Aminopiperidine dihydrochloride ≥97.0% (by titrimetric analysis)
(R)-(-)-3-Aminopiperidine dihydrochloride
Formula: C₅H₁₄Cl₂N₂
MDL Number: MFCD06799458
CAS Number: 334618-23-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (with Total Nitrogen) min. 95.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 206.0 to 210.0 deg-C
Specific rotation [a]20/D -1.0 to -3.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR