1-(1-Boc-piperidino)-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
B5002-1G
B5002-5G
TCB5002-1GEA
270.18
CAD
TCB5002-1G
TCB5002-5G
1-(1-Boc-piperidino)-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
1-(1-Boc-piperidino)-1H-pyrazole-4-boronic acid pinacol ester
Formula:
C₁₉H₃₂BN₃O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD11112131 CAS Number: 877399-74-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 114.0 to 118.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...