5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Chloro-1-(piperidin-4-yl)-1H-benzo[d]imidazol-2(3H)-one, 5-Chloro-1-(4-piperidinyl)-2-benzimidazolinone

C3068-25G C3068-5G
TCC3068-25GEA 279.66 CAD
TCC3068-25G TCC3068-5G
5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone ≥98.0% (by GC, titration analysis)
5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone
Formula: C₁₂H₁₄ClN₃O
MW: 251.72 g/mol
MDL Number: MFCD02093793
CAS Number: 53786-28-0
EINECS: 258-771-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 226.0 to 230.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR