N-Fmoc-β-alanine ≥99.0% (by HPLC)

Supplier: TCI America

Synonyms: Fmoc-β-Ala-OH P, Fmoc-β-Ala-p-alkoxybenzyl alcohol, N-Fmoc-beta-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-beta-alanine

F0882-25G F0882-5G
TCF0882-25GEA 123.24 CAD
TCF0882-25G TCF0882-5G
N-Fmoc-β-alanine ≥99.0% (by HPLC)
N-Fmoc-β-alanine
Formula: C₁₈H₁₇NO₄
MW: 311.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00063328
CAS Number: 35737-10-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR