1-Methylimidazole-2-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Methyl-1H-imidazole-2-carbaldehyde, 1-Methylimidazole-2-carboxaldehyde

M2176-1G M2176-5G
76220-232EA 53.72 CAD
76220-232 76220-234
1-Methylimidazole-2-carbaldehyde ≥98.0% (by GC)
1-Methylimidazole-2-carbaldehyde
Formula: C₅H₆N₂O
Boiling Pt: 74°C
Melting Pt: 39°C
Storage Temperature: Refrigerator
MDL Number: MFCD01321308
CAS Number: 13750-81-7

Order Now

Specification Test Results

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Acid Value max. 5.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR