3-(Isopropoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 3-(Isopropoxycarbonyl)benzeneboronic acid

I1000-1G
TCI1000-1GEA 127.98 CAD
TCI1000-1G
3-(Isopropoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
3-(Isopropoxycarbonyl)phenylboronic acid
Formula: C₁₀H₁₃BO₄
MW: 208.02
MDL Number: MFCD02093047
CAS Number: 342002-80-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR