4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
tert-Butyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 4-(tert-Butoxycarbonyl)phenylboronic acid, pinacol ester, 4-Boc-phenylboronic acid Pinacol ester, 2-(4-tert-Butoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid tert-Butyl ester, 4-(tert-Butoxycarbonyl)phenylboronic acid Pinacol ester
B5132-1G
B5132-5G
TCB5132-1GEA
99.54
CAD
TCB5132-1G
TCB5132-5G
4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester ≥97.0% (by GC, titration analysis)
4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester
Formula:
C₁₇H₂₅BO₄ MW: 304.1890 g/mol |
MDL Number:
MFCD03453055 CAS Number: 850568-72-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...