Methyl-3-methylpicolinate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
M2746-1G
M2746-5G
TCM2746-1GEA
31.6
CAD
TCM2746-1G
TCM2746-5G
Methyl-3-methylpicolinate ≥98.0% (by GC)
Methyl 3-methylpicolinate
Formula:
C₈H₉NO₂ Boiling Pt: 73°C |
MDL Number:
MFCD03789583 CAS Number: 59718-84-2 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...