3-(Trifluoromethoxy)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

T3377-1G T3377-5G
TCT3377-1GEA 75.84 CAD
TCT3377-1G TCT3377-5G
3-(Trifluoromethoxy)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-(Trifluoromethoxy)phenylboronic acid pinacol ester
Formula: C₁₃H₁₆BF₃O₃
MDL Number: MFCD05863919
CAS Number: 262376-31-8

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR