2-Amino-5-chloro-6-methylpyridine ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A2920-1G A2920-5G
TCA2920-1GEA 93.22 CAD
TCA2920-1G TCA2920-5G
2-Amino-5-chloro-6-methylpyridine ≥98.0% (by titrimetric analysis)
2-Amino-5-chloro-6-methylpyridine
Formula: C₆H₇ClN₂
MDL Number: MFCD09453335
CAS Number: 36936-23-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 70.0 to 74.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR