5-Amino-2-bromo-3-methylpyridine ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Amino-2-bromo-3-picoline

A2975-1G A2975-5G
TCA2975-1GEA 36.34 CAD
TCA2975-1G TCA2975-5G
5-Amino-2-bromo-3-methylpyridine ≥95.0% (by GC, titration analysis)
5-Amino-2-bromo-3-methylpyridine
Formula: C₆H₇BrN₂
MDL Number: MFCD03095085
CAS Number: 38186-83-3

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 98.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR