4-Bromo-4'-methylbiphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Bromo-4'-methyl-1,1'-biphenyl

B4804-1G B4804-5G
TCB4804-1GEA 347.6 CAD
TCB4804-1G TCB4804-5G
4-Bromo-4'-methylbiphenyl ≥98.0% (by GC)
4-Bromo-4'-methylbiphenyl
Formula: C₁₃H₁₁Br
MW: 247.14 g/mol
Boiling Pt: 315°C
MDL Number: MFCD00466236
CAS Number: 50670-49-0

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 131.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR