4-Cyano-4'-octylbiphenyl ≥98.0% (by GC)

Supplier: TCI America
Warning

C3156-25G C3156-5G
TCC3156-25GEA 913.24 CAD
TCC3156-25G TCC3156-5G
4-Cyano-4'-octylbiphenyl ≥98.0% (by GC)
4-Cyano-4'-octylbiphenyl
Formula: C₂₁H₂₅N
Boiling Pt: 218°C
MDL Number: MFCD00075146
CAS Number: 52709-84-9
EINECS: 258-120-6

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR