Ethyl thiophene-3-carboxylate ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl 3-Thiophenecarboxylate, 3-Thiophenecarboxylic acid ethyl ester

E1082-1G E1082-5G
TCE1082-1GEA 50.56 CAD
TCE1082-1G TCE1082-5G
Ethyl thiophene-3-carboxylate ≥95.0% (by GC)
Ethyl thiophene-3-carboxylate
Formula: C₇H₈O₂S
MW: 156.20 g/mol
Boiling Pt: 80°C
MDL Number: MFCD02179226
CAS Number: 5751-80-4
EINECS: 227-275-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR