Ethyl-5-bromothiophene-2-carboxylate ≥98.0% (by GC)
Supplier: TCI America
E1111-1G
E1111-5G
TCE1111-1GEA
42.66
CAD
TCE1111-1G
TCE1111-5G
Ethyl-5-bromothiophene-2-carboxylate ≥98.0% (by GC)
Ethyl 5-bromothiophene-2-carboxylate
Formula:
C₇H₇BrO₂S Boiling Pt: 175°C |
MDL Number:
MFCD02683089 CAS Number: 5751-83-7 |
Specification Test Results
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...