2-Methoxy-4-methyl-5-nitropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

M2652-1G M2652-5G
TCM2652-1GEA 75.84 CAD
TCM2652-1G TCM2652-5G
2-Methoxy-4-methyl-5-nitropyridine ≥98.0% (by GC)
2-Methoxy-4-methyl-5-nitropyridine
Formula: C₇H₈N₂O₃
MDL Number: MFCD03095075
CAS Number: 6635-90-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR