Ethyl-4-hydroxyquinoline-3-carboxylate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
E1142-1G
E1142-5G
TCE1142-1GEA
76
CAD
TCE1142-1G
TCE1142-5G
Ethyl-4-hydroxyquinoline-3-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl 4-hydroxyquinoline-3-carboxylate
Formula:
C₁₂H₁₁NO₃ Melting Pt: 271°C |
MDL Number:
MFCD00173406 CAS Number: 26892-90-0 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...