4-Cyano-4'-hexyloxybiphenyl ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
4'-Hexyloxy-4-biphenylcarbonitrile, 4'-Hexyloxy-[1,1'-biphenyl]-4-carbonitrile, 6OCB, 4'-Cyano-4-hexyloxybiphenyl
C3155-25G
C3155-5G
TCC3155-25GEA
1019.89
CAD
TCC3155-25G
TCC3155-5G
4-Cyano-4'-hexyloxybiphenyl ≥98.0% (by GC)
4-Cyano-4'-hexyloxybiphenyl
Formula:
C₁₉H₂₁NO Melting Pt: 58°C |
MDL Number:
MFCD00134145 CAS Number: 41424-11-7 EINECS: 255-358-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...