1-(4-Cyanophenyl)piperazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-(Piperazin-1-yl)benzonitrile, 4-(1-Piperazinyl)benzonitrile, 1-(4-Cyanophenyl)piperazine

P2449-1G P2449-5G
TCP2449-1GEA 208.56 CAD
TCP2449-1G TCP2449-5G
1-(4-Cyanophenyl)piperazine ≥98.0% (by GC, titration analysis)
1-(4-Cyanophenyl)piperazine
Formula: C₁₁H₁₃N₃
MDL Number: MFCD01320890
CAS Number: 68104-63-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR