4-[4-(Trifluoromethoxy)phenoxy]piperidine ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-(4-Trifluoromethoxyphenoxy)piperidine

T3349-1G T3349-5G
TCT3349-1GEA 77.42 CAD
TCT3349-1G TCT3349-5G
4-[4-(Trifluoromethoxy)phenoxy]piperidine ≥97.0% (by GC, titration analysis)
4-[4-(Trifluoromethoxy)phenoxy]piperidine
Formula: C₁₂H₁₄F₃NO₂
MW: 261.25 g/mol
MDL Number: MFCD06656166
CAS Number: 287952-67-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 74.0 to 78.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR