Methyl-4-amino-3-chlorobenzoate ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
M2698-1G
M2698-5G
TCM2698-1GEA
74.42
CAD
TCM2698-1G
TCM2698-5G
Methyl-4-amino-3-chlorobenzoate ≥97.0% (by GC)
Methyl 4-amino-3-chlorobenzoate
Formula:
C₈H₈ClNO₂ |
MDL Number:
MFCD01320686 CAS Number: 84228-44-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 107.0 to 111.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...