Methyl-5-amino-2-chlorobenzoate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

M2812-1G M2812-5G
TCM2812-1GEA 41.08 CAD
TCM2812-1G TCM2812-5G
Methyl-5-amino-2-chlorobenzoate ≥98.0% (by titrimetric analysis)
Methyl 5-amino-2-chlorobenzoate
Formula: C₈H₈ClNO₂
Boiling Pt: 160°C
Storage Temperature: Refrigerator
MDL Number: MFCD07800315
CAS Number: 42122-75-8

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to lump
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR