1,4-Difluoro-2-methyl-5-nitrobenzene ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D4806-1G
D4806-5G
76219-704EA
109.02
CAD
76219-704
76219-706
1,4-Difluoro-2-methyl-5-nitrobenzene ≥98.0% (by GC)
1,4-Difluoro-2-methyl-5-nitrobenzene
Formula:
C₇H₅F₂NO₂ |
MDL Number:
MFCD06200905 CAS Number: 141412-60-4 |
Specification Test Results
Appearance | White to Yellow to Green powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...