1-Bromo-4-chloro-3-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

B4374-25G B4374-5G
TCB4374-25GEA 193.39 CAD
TCB4374-25G TCB4374-5G
1-Bromo-4-chloro-3-nitrobenzene ≥98.0% (by GC)
1-Bromo-4-chloro-3-nitrobenzene
Formula: C₆H₃BrClNO₂
MDL Number: MFCD05863216
CAS Number: 16588-24-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 74.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR