3,6-Dibromopyridazine ≥98.0% (by GC)

Supplier: TCI America
Warning

D5466-1G D5466-5G
TCD5466-1GEA 52.14 CAD
TCD5466-1G TCD5466-5G
3,6-Dibromopyridazine ≥98.0% (by GC)
3,6-Dibromopyridazine
Formula: C₄H₂Br₂N₂
MDL Number: MFCD00233947
CAS Number: 17973-86-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR