[5-Phenyl-3-isoxazolyl]methanol ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(5-Phenylisoxazol-3-yl)methanol, 3-Hydroxymethyl-5-phenylisoxazole
P2386-1G
P2386-5G
76220-420EA
252.8
CAD
76220-420
76220-422
[5-Phenyl-3-isoxazolyl]methanol ≥98.0% (by GC)
[5-Phenyl-3-isoxazolyl]methanol
Formula:
C₁₀H₉NO₂ MW: 175.1840 g/mol |
MDL Number:
MFCD01444104 CAS Number: 1619-37-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...