Ethyl-2-chloro-6-methylisonicotinate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Ethyl 2-chloro-6-methylpyridine-4-carboxylate, Ethyl 2-chloro-6-methylisonicotinate

E1262-1G E1262-5G
TCE1262-1GEA 74.26 CAD
TCE1262-1G TCE1262-5G
Ethyl-2-chloro-6-methylisonicotinate ≥98.0% (by GC)
Ethyl 2-chloro-6-methylisonicotinate
Formula: C₉H₁₀ClNO₂
MW: 199.64 g/mol
Boiling Pt: 107°C
MDL Number: MFCD09842151
CAS Number: 3998-88-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR