2-Amino-5-bromo-4-methyl-3-nitropyridine ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A2988-1G A2988-5G
TCA2988-1GEA 107.44 CAD
TCA2988-1G TCA2988-5G
2-Amino-5-bromo-4-methyl-3-nitropyridine ≥98.0% (by titrimetric analysis)
2-Amino-5-bromo-4-methyl-3-nitropyridine
Formula: C₆H₆BrN₃O₂
MW: 232.0400 g/mol
MDL Number: MFCD00092197
CAS Number: 100367-40-6

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 168.0 to 172.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR