4-Benzyloxy-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
B4638-1G
B4638-200MG
TCB4638-1GEA
194.34
CAD
TCB4638-1G
TCB4638-200MG
4-Benzyloxy-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
4-Benzyloxy-2-fluorophenylboronic acid
Formula:
C₁₃H₁₂BFO₃ |
MDL Number:
MFCD02093064 CAS Number: 166744-78-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 108.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...