2-(Benzyloxy)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
B4590-1G
B4590-5G
TCB4590-1GEA
61.62
CAD
TCB4590-1G
TCB4590-5G
2-(Benzyloxy)phenylboronic acid (contains varying amounts of Anhydride)
2-(Benzyloxy)phenylboronic acid
Formula:
C₁₃H₁₃BO₃ Melting Pt: 110°C |
MDL Number:
MFCD01632206 CAS Number: 190661-29-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 109.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...