4-Fluoro-3-nitrobenzenesulfonamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

F1172-1G F1172-5G
TCF1172-1GEA 186.44 CAD
TCF1172-1G TCF1172-5G
4-Fluoro-3-nitrobenzenesulfonamide ≥98.0% (by HPLC, total nitrogen)
4-Fluoro-3-nitrobenzenesulphonamide
Formula: C₆H₅FN₂O₄S
MDL Number: MFCD08703185
CAS Number: 406233-31-6

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 137.0 to 141.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR