5-Cyano-2-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

C3227-1G C3227-5G
TCC3227-1GEA 216.46 CAD
TCC3227-1G TCC3227-5G
5-Cyano-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
5-Cyano-2-fluorophenylboronic acid
Formula: C₇H₅BFNO₂
MDL Number: MFCD03095147
CAS Number: 468718-30-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR