4-Amino-2-methoxybenzonitrile ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
A2785-1G
A2785-5G
TCA2785-1GEA
249.64
CAD
TCA2785-1G
TCA2785-5G
4-Amino-2-methoxybenzonitrile ≥97.0% (by GC, titration analysis)
4-Amino-2-methoxybenzonitrile
Formula:
C₈H₈N₂O |
MDL Number:
MFCD09027303 CAS Number: 7251-09-4 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...